Learn More
Medchemexpress LLC GUCY2C/Guanylyl cycl 10ug | 10ug
SDP
Catalog No. 5000194216
Encompass_Preferred
GUCY2C a vital guanylyl cyclase catalyzes the synthesis of cyclic cGMP from GTP GUCY2C/Guanylyl cyclase C Protein Mouse (HEK293 His) is the recombinant mouse-derived GUCY2C/Guanylyl cyclase C protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-94216 Supplier Medchemexpress LLC Supplier No. HYP7874210UG
GUCY2C a vital guanylyl cyclase catalyzes the synthesis of cyclic cGMP from GTP GUCY2C/Guanylyl cyclase C Protein Mouse (HEK293 His) is the recombinant mouse-derived GUCY2C/Guanylyl cyclase C protein expressed by HEK293 with C-6 His labeled tag