Learn More
Medchemexpress LLC GUCY2C/Guanylyl cycl 10ug
SDP
Catalog No. 5000196812
Encompass_Preferred
Recombinant rat Guanylyl Cyclase C (GUCY2C) produced in HEK293 cells with N-terminal His and C-terminal Myc tags. Supplied in small pack sizes (10 μg available) for use in biochemical and cell-based assays to study receptor activation, cGMP production, and toxin-receptor interactions. For research use only.
  • Expressed in HEK293 cells for mammalian-like post-translational modifications.
  • Includes N-terminal His tag and C-terminal Myc tag for purification and detection.
  • Suitable for studies of receptor activation and cGMP signaling.
  • Available in small 10 μg pack sizes for assay development.
  • Provided for research use only; not for human or diagnostic use.
Catalog No. 50-001-96812 Supplier Medchemexpress LLC Supplier No. HYP7166610UG
Recombinant rat Guanylyl Cyclase C (GUCY2C) produced in HEK293 cells with N-terminal His and C-terminal Myc tags. Supplied in small pack sizes (10 μg available) for use in biochemical and cell-based assays to study receptor activation, cGMP production, and toxin-receptor interactions. For research use only.
  • Expressed in HEK293 cells for mammalian-like post-translational modifications.
  • Includes N-terminal His tag and C-terminal Myc tag for purification and detection.
  • Suitable for studies of receptor activation and cGMP signaling.
  • Available in small 10 μg pack sizes for assay development.
  • Provided for research use only; not for human or diagnostic use.