Learn More
ABclonal Technology GUCY2F Rabbit pAb
SDP
Supplier:  ABclonal Technology A142421000UL
Encompass_Preferred
The protein encoded by this gene is a guanylyl cyclase found predominantly in photoreceptors in the retina. The encoded protein is thought to be involved in resynthesis of cGMP after light activation of the visual signal transduction cascade, allowing a return to the dark state. This protein is a single-pass type I membrane protein. Defects in this gene may be a cause of X-linked retinitis pigmentosa.
Catalog No. 50-001-09039