Learn More
Aviva Systems Biology Corp Guf1 Antibody - C-terminal region (ARP57564_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57564_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Guf1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 4631409J12, AA407526, EF-4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-286-333