Learn More
Selleck Chemical LLC Guggulsterone E&Z S3792-50mg
SDP
Catalog No. 503109044
Encompass_Preferred
Guggulsterone is one of the active constituent of Commiphora mukul It occurs in two isomeric forms namely Z-GS and E-GS Guggulsterone act as antagonist ligands for the bile acid receptor farnesoid X receptor and as active ingredients responsible for the hypolipidemic activity
Catalog No. 50-310-9044 Supplier Selleck Chemical LLC Supplier No. S379250MG
Guggulsterone is one of the active constituent of Commiphora mukul It occurs in two isomeric forms namely Z-GS and E-GS Guggulsterone act as antagonist ligands for the bile acid receptor farnesoid X receptor and as active ingredients responsible for the hypolipidemic activity