Learn More
Takara Bio Guide-it™ Complete sgRNA Screening System, 50 Rxns
SDP
Catalog No. NC1659911
Encompass
The Guide-it Complete sgRNA Screening System includes everything needed for the simple production, cleanup, and evaluation of single guide RNAs (sgRNAs) for CRISPR/Cas9 studies, including PCR reagents for amplifying your genomic target and recombinant Cas9 for in vitro analysis of the transcribed sgRNA.
Catalog No. NC1659911 Supplier Takara Bio Supplier No. 632636
The Guide-it Complete sgRNA Screening System includes everything needed for the simple production, cleanup, and evaluation of single guide RNAs (sgRNAs) for CRISPR/Cas9 studies, including PCR reagents for amplifying your genomic target and recombinant Cas9 for in vitro analysis of the transcribed sgRNA.