Learn More
Takara Bio Guide-it Long ssDNA Production System v2 | 50 rxns
SDP
Catalog No. NC1816249
Encompass
The Guide-it Long ssDNA Production System v2 enables on-demand production of long single-stranded DNA (ssDNA) up to 5,000 nt in length for use as a repair template in knockin experiments involving CRISPR/Cas9 technology or other genome editing tools. This kit provides a simple and fast method for converting a dsDNA PCR product of any sequence into ssDNA via selective digestion of either the sense or the antisense strand. Each kit allows for preparation of 50 ssDNA templates.
Catalog No. NC1816249 Supplier Takara Bio Supplier No. 632666
The Guide-it Long ssDNA Production System v2 enables on-demand production of long single-stranded DNA (ssDNA) up to 5,000 nt in length for use as a repair template in knockin experiments involving CRISPR/Cas9 technology or other genome editing tools. This kit provides a simple and fast method for converting a dsDNA PCR product of any sequence into ssDNA via selective digestion of either the sense or the antisense strand. Each kit allows for preparation of 50 ssDNA templates.