Learn More
Thermo Scientific Flow Tubes | Stainless Steel | 50Set
Catalog No. NC2131633 Shop All Thermo Scientific Products
Encompass
High purity, Customizable length, Suitable for ELEMENT GD GD-MS system, Intended for single use to avoid cross-contamination
This item is not returnable. View return policy
Catalog No. NC2131633 Supplier Thermo Scientific Supplier No. 1194561
High purity, Customizable length, Suitable for ELEMENT GD GD-MS system, Intended for single use to avoid cross-contamination