Learn More
Stoelting™ Guinea Pig Adaptor Kit
SDP
Catalog No. 10000149 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Adaptor kit
Catalog No. Model
10-000-149 Adaptor kit
1 options
Catalog No. 10-000-149 Supplier Stoelting™ Supplier No. 51622
 Restricted Item
Add to Cart
Add to Cart
Includes the 45° ear bars required for guinea pigs.
  • Nose clamp reaches further back behind the incisors than on the rat adaptors
  • Capable of both vertical and A-P travel
Specifications
For Use With (Application) Neuroscience
Animal Guinea Pig
Model Adaptor kit