Learn More
Biomatik Corporation Guinea pig Albumin (ALB) ELISA Kit
Supplier:  Biomatik Corporation EKU02213
Encompass_Preferred
Guinea pig Albumin (ALB) ELISA Kit for detecting ALB in serum, plasma, other biological fluids. Species Reactivity: Guinea pig (Cavia porcellus). Detection Range: 1.23-100ug/ml. Sensitivity: 0.43ug/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: H0V003. Target Synonyms: Serum albumin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5097