Learn More
Immunology Consultants Guinea Pig C3 ELISA Kit
SDP
Supplier:  Immunology Consultants E60C3
Encompass_Preferred
The Guinea Pig C3 test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring C3 in guinea pig biological samples.
Catalog No. 50-240-6650