Learn More
Sino Biological Anti-CD1E Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107572T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Guinea pig CD1E, and purified by antigen affinity chromatography.
Catalog No. 50-241-2411