Learn More
Biomatik Corporation Guinea pig Gastrin (GT) ELISA Kit
Supplier:  Biomatik Corporation EKU04345
Encompass_Preferred
Guinea pig Gastrin (GT) ELISA Kit for detecting GT in serum, plasma, other biological fluids. Species Reactivity: Guinea pig (Cavia porcellus). Detection Range: 12.35-1000pg/ml. Sensitivity: 4.35pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P06885. Target Synonyms: GAST; GAS; Gastrin component I; Big gastrin; Gastrin component II; Gastrin component III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5658