Learn More
Bio Serv GUINEA PIG HUT CERTIFIED
SDP
Supplier:  Bio Serv K3261
Encompass
Guinea Pig Hut, CertifiedCertified (Contaminant Screened)Made of polycarbonateProvides a comfortable hiding placeTransparent material allows for ease of visibilityStacks easily for storageCagewasher safe onlyPlease Note: There is a protective layer on both sides of the polycarbonate products. One layer is white and the other side is clear and not obvious to the end user. Please assure both sides of the protective layers have been removed before the products are put into use. qty 1-49
Catalog No. NC3252418