Learn More
American Research Products Inc Guinea pig Immunoglobulin E I
Catalog No. 5000329765
Encompass
Guinea pig Immunoglobulin E (IgE) ELISA kit from CUSABIO Detection Range 93 75 ng/mL-6000 ng/mL Sensitivity 23 4 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29765 Supplier American Research Products Inc Supplier No. CSBE12102GP
Guinea pig Immunoglobulin E (IgE) ELISA kit from CUSABIO Detection Range 93 75 ng/mL-6000 ng/mL Sensitivity 23 4 ng/mL Sample serum plasma tissue homogenates Method Sandwich