Learn More
American Research Products Inc Guinea pig Immunoglobulin G I
Catalog No. 5000326201
Encompass
Guinea pig Immunoglobulin G IgG ELISA Kit from CUSABIO Detection Range 62 5 g/mL-1000 g/mL Sensitivity 62 5 g/mL Sample serum plasma Method Competitive
Catalog No. 50-003-26201 Supplier American Research Products Inc Supplier No. CSBE10125GU
Guinea pig Immunoglobulin G IgG ELISA Kit from CUSABIO Detection Range 62 5 g/mL-1000 g/mL Sensitivity 62 5 g/mL Sample serum plasma Method Competitive