Learn More
Biomatik Corporation Guinea pig Insulin Like Growth Factor 1 (IGF1) ELISA Kit
Supplier:  Biomatik Corporation EKU04966
Encompass_Preferred
Guinea pig Insulin Like Growth Factor 1 (IGF1) ELISA Kit for detecting IGF1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Guinea pig (Cavia porcellus). Detection Range: 78-5,000pg/ml. Sensitivity: 31pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P17647. Gene ID: 100135551. Target Synonyms: IGFI; IGF1A; IBP1; MGF; Somatomedin C; Mechano Growth Gactor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5699