Learn More
American Research Products Inc Guinea pig Interferon y IFN y
Catalog No. 5000326501
Encompass
Guinea pig Interferon y IFN-y ELISA Kit from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 7 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26501 Supplier American Research Products Inc Supplier No. CSBE06763GU
Guinea pig Interferon y IFN-y ELISA Kit from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 7 pg/mL Sample serum plasma tissue homogenates Method Sandwich