Learn More
American Research Products Inc Guinea pig Interleukin 1 beta
Catalog No. 5000327861
Encompass
Guinea pig Interleukin-1 beta (IL1B) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27861 Supplier American Research Products Inc Supplier No. CSBEL011614GU
Guinea pig Interleukin-1 beta (IL1B) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich