Learn More
American Research Products Inc Guinea pig interleukin 10 IL
Catalog No. 5000329014
Encompass
Guinea pig interleukin 10 IL-10 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29014 Supplier American Research Products Inc Supplier No. CSBE12915GP
Guinea pig interleukin 10 IL-10 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich