Learn More
Biomatik Corporation Guinea pig Interleukin 12B (IL12B) ELISA Kit
Supplier:  Biomatik Corporation EKU05210
Encompass_Preferred
Guinea pig Interleukin 12B (IL12B) ELISA Kit for detecting IL12B in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Guinea pig (Cavia porcellus). Detection Range: 7.81-500pg/ml. Sensitivity: 3.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q924V5. Gene ID: 100135554. Target Synonyms: p40; IL12-B; CLMF2; NKSF2; Natural Killer Cell Stimulatory Factor 2; Cytotoxic Lymphocyte Maturation Factor 2; Interleukin 23, P40 Subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5622