Learn More
American Research Products Inc Guinea pig interleukin 2 IL 2
Catalog No. 5000329013
Encompass
Guinea pig interleukin 2 IL-2 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29013 Supplier American Research Products Inc Supplier No. CSBE12914GP
Guinea pig interleukin 2 IL-2 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich