Learn More
American Research Products Inc Guinea pig interleukin 4 IL 4
Catalog No. 5000329176
Encompass
Guinea pig interleukin 4 IL-4 ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29176 Supplier American Research Products Inc Supplier No. CSBE10126GU
Guinea pig interleukin 4 IL-4 ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich