Learn More
American Research Products Inc Guinea pig interleukin 5 IL 5
Catalog No. 5000329158
Encompass
Guinea pig interleukin 5 IL-5 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29158 Supplier American Research Products Inc Supplier No. CSBE12064GP
Guinea pig interleukin 5 IL-5 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich