Learn More
American Research Products Inc Guinea pig Interleukin 8 IL 8
Catalog No. 5000329043
Encompass
Guinea pig Interleukin 8 IL-8 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29043 Supplier American Research Products Inc Supplier No. CSBE13096GP
Guinea pig Interleukin 8 IL-8 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich