Learn More
American Research Products Inc guinea pig matriX metalloprote
Catalog No. 5000326306
Encompass
guinea pig matriX metalloproteinase 9 MMP-9 ELISA Kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/ mL Sensitivity 0 78 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26306 Supplier American Research Products Inc Supplier No. CSBE06765GU
guinea pig matriX metalloproteinase 9 MMP-9 ELISA Kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/ mL Sensitivity 0 78 ng/mL Sample serum plasma tissue homogenates Method Sandwich