Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Guinea Pig Mineral Mix

Catalog No. MP296028550 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 kg
10 kg
50 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP296028550 50 kg
MP296028502 2 kg
MP296028510 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP296028550 Supplier MP Biomedicals™ Supplier No. 0296028550
Add to Cart
Add to Cart

Provides complete minerals for normal growth and development of the guinea pig when added at a level of 6.75% of the diet.

  • Salt Mixture
  • For guinea pig diet
TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
For Use With (Application) Guinea pig diet
Quantity 50 kg
Storage Requirements Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.