Learn More
Biomatik Corporation Guinea pig Monocyte Chemotactic Protein 1 (MCP1) ELISA Kit
Supplier:  Biomatik Corporation EKU11290
Encompass_Preferred
Guinea pig Monocyte Chemotactic Protein 1 (MCP1) ELISA Kit for detecting MCP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Guinea pig (Cavia porcellus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q08782. Gene ID: 100135494. Target Synonyms: CCL2; GDCF2; HC11; HSMCR30; MCAF; MCP1; SCYA2; SMC-CF; Chemokine C-C-Motif Ligand 2; Monocyte Chemotactic And Activating Factor; Monocyte Secretory Protein JE. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5275