Learn More
Biomatik Corporation Guinea pig Neuropeptide Y (NPY) ELISA Kit
Supplier:  Biomatik Corporation EKU11578
Encompass_Preferred
Guinea pig Neuropeptide Y (NPY) ELISA Kit for detecting NPY in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Guinea pig (Cavia porcellus). Detection Range: 24.69-2,000pg/ml. Sensitivity: 8.87pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P68008. Target Synonyms: NP-Y; PYY4; CPON; Pro-neuropeptide Y; Neuropeptide tyrosine; C-flanking peptide of NPY. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6559