Learn More
Innovative Research Inc Guinea Pig Red Blood Cells 25ml
Catalog No. 502034854
Encompass_Preferred
Guinea Pig Red Blood Cells from Innovative Research are manufactured using whole blood collections from Guinea Pigs located in the USA. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.
Catalog No. 50-203-4854 Supplier Innovative Research Inc Supplier No. IGPRBC25ML
Guinea Pig Red Blood Cells from Innovative Research are manufactured using whole blood collections from Guinea Pigs located in the USA. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.