Learn More
Biomatik Corporation Guinea pig Substance P (SP) CLIA Kit
Supplier:  Biomatik Corporation EKU08614
Encompass_Preferred
Guinea pig Substance P (SP) CLIA Kit for detecting SP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Guinea pig (Cavia porcellus). Detection Range: 11.72-3,000pg/ml. Sensitivity: 5.12pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P67932. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8653