Learn More
American Research Products Inc Guinea pig tri iodothyronine
Catalog No. 5000328024
Encompass
Guinea pig tri-iodothyronine (T3) ELISA kit from CUSABIO Detection Range 0 5 ng/mL-8 ng/mL Sensitivity 0 25 ng/mL Sample serum plasma Method Competitive
Catalog No. 50-003-28024 Supplier American Research Products Inc Supplier No. CSBEQ027510GU
Guinea pig tri-iodothyronine (T3) ELISA kit from CUSABIO Detection Range 0 5 ng/mL-8 ng/mL Sensitivity 0 25 ng/mL Sample serum plasma Method Competitive