Learn More
American Research Products Inc Guinea pig Tumor necrosis fact
Catalog No. 5000329927
Encompass
Guinea pig Tumor necrosis factor a TNF-a ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29927 Supplier American Research Products Inc Supplier No. CSBE06772GU
Guinea pig Tumor necrosis factor a TNF-a ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich