Learn More
Biomatik Corporation Guinea pig Vasoactive Intestinal Peptide (VIP) ELISA Kit
Supplier:  Biomatik Corporation EKU08112
Encompass_Preferred
Guinea pig Vasoactive Intestinal Peptide (VIP) ELISA Kit for detecting VIP in serum, plasma, other biological fluids. Species Reactivity: Guinea pig (Cavia porcellus). Detection Range: 6.17-500pg/ml. Sensitivity: 2.52pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P04566. Target Synonyms: PHM27; Intestinal peptide PHV-42; Peptide histidine valine 42; Peptide histidine methioninamide 27. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4476