Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antibodies Online Guinea Pig anti-Rabbit IgG, Polyclonal, 200 μL, 1.2 mg/mL, Preadsorbed
SDP

Catalog No. NC2013818
Encompass

This Guinea Pig anti-Rabbit IgG (Heavy & Light Chain) antibody (ABIN101961) is a preadsorbed polyclonal antibody developed in guinea pigs. It specifically targets the heavy and light chains of rabbit IgG with no observed cross-reactivity against human, mouse, or goat serum proteins. This antibody is validated for a variety of scientific applications, including ELISA, Western Blot, immunohistochemistry (IHC), CUT&RUN, and CUT&Tag.

  • Targets heavy and light chains of rabbit IgG
  • No cross-reactivity against human, mouse, or goat serum proteins
  • Validated for ELISA, Western Blot, IHC, CUT&RUN, and CUT&Tag applications
  • Preadsorbed polyclonal antibody
  • Recommended concentration for CUT&RUN/CUT&Tag: 1.2 mg/mL
  • Volume: 200 μL
  • Short-term storage: 4 °C
  • Long-term storage: -20 °C or below (aliquoted)
  • Preservative: Sodium azide

Catalog No. NC2013818 Supplier Antibodies Online Supplier No. ABIN101961
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Guinea Pig anti-Rabbit IgG (Heavy & Light Chain) antibody (ABIN101961) is a preadsorbed polyclonal antibody developed in guinea pigs. It specifically targets the heavy and light chains of rabbit IgG with no observed cross-reactivity against human, mouse, or goat serum proteins. This antibody is validated for a variety of scientific applications, including ELISA, Western Blot, immunohistochemistry (IHC), CUT&RUN, and CUT&Tag.

  • Targets heavy and light chains of rabbit IgG
  • No cross-reactivity against human, mouse, or goat serum proteins
  • Validated for ELISA, Western Blot, IHC, CUT&RUN, and CUT&Tag applications
  • Preadsorbed polyclonal antibody
  • Recommended concentration for CUT&RUN/CUT&Tag: 1.2 mg/mL
  • Volume: 200 μL
  • Short-term storage: 4 °C
  • Long-term storage: -20 °C or below (aliquoted)
  • Preservative: Sodium azide

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.