Learn More
Aviva Systems Biology Corp GULP1 Antibody - N-terminal region (ARP65446_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65446_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GULP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human GULP1. Alternate names: CED-6, CED6, FLJ31156, GULPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-288-499