Learn More
Medchemexpress LLC Guluronic acid sodium | 32510-76-2 | 10 MG
SDP
Catalog No. 5000419948
Encompass_Preferred
Guluronic acid sodium (G2013) is an orally active oxidative stress modulator and anti-inflammatory agent. It down-regulates pro-inflammatory and oxidative stress genes (e.g., TLR4, NF-κB, iNOS) and inhibits COX-2, MMPs, and VEGF. It also up-regulates immunoregulatory genes SHIP1 and SOCS1, suppressing cancer-related inflammation, angiogenesis, cell adhesion, and metastasis. This compound shows a favorable safety profile and has been investigated in multiple sclerosis, ankylosing spondylitis, and breast cancer research.
  • Orally active oxidative stress modulator
  • Anti-inflammatory agent
  • Down-regulates pro-inflammatory and oxidative stress genes
  • Inhibits COX-2, MMPs, and VEGF
  • Suppresses cancer-related inflammation, angiogenesis, and metastasis
  • Reduces immunosuppressive cell accumulation
  • Investigated in multiple sclerosis and cancer research
Catalog No. 50-004-19948 Supplier Medchemexpress LLC Supplier No. HYN7700A10MG
Add to Cart
Add to Cart
Guluronic acid sodium (G2013) is an orally active oxidative stress modulator and anti-inflammatory agent. It down-regulates pro-inflammatory and oxidative stress genes (e.g., TLR4, NF-κB, iNOS) and inhibits COX-2, MMPs, and VEGF. It also up-regulates immunoregulatory genes SHIP1 and SOCS1, suppressing cancer-related inflammation, angiogenesis, cell adhesion, and metastasis. This compound shows a favorable safety profile and has been investigated in multiple sclerosis, ankylosing spondylitis, and breast cancer research.
  • Orally active oxidative stress modulator
  • Anti-inflammatory agent
  • Down-regulates pro-inflammatory and oxidative stress genes
  • Inhibits COX-2, MMPs, and VEGF
  • Suppresses cancer-related inflammation, angiogenesis, and metastasis
  • Reduces immunosuppressive cell accumulation
  • Investigated in multiple sclerosis and cancer research