Learn More
Medchemexpress LLC Guluronic acid sodium | 32510-76-2 | 5 MG
SDP
Catalog No. 5000418845
Encompass_Preferred
Guluronic acid sodium is an orally active oxidative stress modulator and anti-inflammatory agent. It down-regulates pro-inflammatory and oxidative stress-related genes while inhibiting enzymes like COX-2 and MMPs. Also up-regulating immunoregulatory genes, it suppresses cancer-related inflammation, tumor angiogenesis, cell adhesion, and metastasis. This compound is researched for multiple sclerosis, ankylosing spondylitis, breast cancer, and other inflammatory diseases.
  • Acts as an oxidative stress modulator and anti-inflammatory agent.
  • Down-regulates pro-inflammatory and oxidative stress-related genes.
  • Inhibits COX-2, MMPs, and VEGF activities.
  • Suppresses cancer-related inflammation, angiogenesis, and metastasis.
  • Reduces immunosuppressive cell accumulation.
  • Orally active.
  • For research use only.
  • Appearance: solid, white to light yellow.
Catalog No. 50-004-18845 Supplier Medchemexpress LLC Supplier No. HYN7700A5MG
Add to Cart
Add to Cart
Guluronic acid sodium is an orally active oxidative stress modulator and anti-inflammatory agent. It down-regulates pro-inflammatory and oxidative stress-related genes while inhibiting enzymes like COX-2 and MMPs. Also up-regulating immunoregulatory genes, it suppresses cancer-related inflammation, tumor angiogenesis, cell adhesion, and metastasis. This compound is researched for multiple sclerosis, ankylosing spondylitis, breast cancer, and other inflammatory diseases.
  • Acts as an oxidative stress modulator and anti-inflammatory agent.
  • Down-regulates pro-inflammatory and oxidative stress-related genes.
  • Inhibits COX-2, MMPs, and VEGF activities.
  • Suppresses cancer-related inflammation, angiogenesis, and metastasis.
  • Reduces immunosuppressive cell accumulation.
  • Orally active.
  • For research use only.
  • Appearance: solid, white to light yellow.