Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gum arabic | 9000-01-5 | 10 G
SDP

Catalog No. 5000418936
Encompass_Preferred

Gum arabic is an orally active complex branched polysaccharide isolated from the *Acacia senegal* tree. It functions as a stabilizer and thickener, and is utilized in research for brain tumor imaging. This compound has been shown to upregulate maturation markers, promote ERK1/2 phosphorylation, and inhibit apoptosis, offering a range of biological activities including antimalarial, hepatoprotective, renal, cardiovascular protective, and anti-obesity effects.

  • Appearance: solid, off-white to light yellow
  • Molecular weight: 200-300
  • Structure classification: saccharides, polysaccharides
  • Isolated from *Acacia senegal* tree
  • Upregulates maturation markers (CD86, CD40, CD54)
  • Promotes ERK1/2 phosphorylation
  • Inhibits apoptosis
  • Exhibits antimalarial effects
  • Offers hepatoprotective, renal, and cardiovascular benefits
  • Improves obesity

Catalog No. 50-004-18936 Supplier Medchemexpress LLC Supplier No. HYN666410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gum arabic is an orally active complex branched polysaccharide isolated from the *Acacia senegal* tree. It functions as a stabilizer and thickener, and is utilized in research for brain tumor imaging. This compound has been shown to upregulate maturation markers, promote ERK1/2 phosphorylation, and inhibit apoptosis, offering a range of biological activities including antimalarial, hepatoprotective, renal, cardiovascular protective, and anti-obesity effects.

  • Appearance: solid, off-white to light yellow
  • Molecular weight: 200-300
  • Structure classification: saccharides, polysaccharides
  • Isolated from *Acacia senegal* tree
  • Upregulates maturation markers (CD86, CD40, CD54)
  • Promotes ERK1/2 phosphorylation
  • Inhibits apoptosis
  • Exhibits antimalarial effects
  • Offers hepatoprotective, renal, and cardiovascular benefits
  • Improves obesity

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.