Learn More
Sigma Aldrich Fine Chemicals Biosciences Gum arabic from acacia tre1KG
Catalog No. 502897579
Encompass
Gum arabic from acacia tree is extracted from the branches of Acacia senegalnbspandnbspAcacia seyalnbsptrees. It is an edible dried gummy exudate. Gum Arabic has high solubility and is used in food industry as a stabilizer emulsifier flavouring agent thickener and surface-finishing agent. It initiates turbidity or hinders sugar crystallization. Gum arabicnbspinhibits color pigmentation and protein precipitation in wine production.
Catalog No. 50-289-7579 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G97521KG
Gum arabic from acacia tree is extracted from the branches of Acacia senegalnbspandnbspAcacia seyalnbsptrees. It is an edible dried gummy exudate. Gum Arabic has high solubility and is used in food industry as a stabilizer emulsifier flavouring agent thickener and surface-finishing agent. It initiates turbidity or hinders sugar crystallization. Gum arabicnbspinhibits color pigmentation and protein precipitation in wine production.