Learn More
Medchemexpress LLC Gum ghatti | 9000-28-6 | ≈12,000 g/mol | 50 G
SDP
Catalog No. 5000283838
Encompass_Preferred
Gum ghatti is a natural exudate polysaccharide supplied as a dry powder for research use. It serves as a biochemical reagent and excipient for assays, formulations, and other laboratory applications requiring a water-soluble gum. The material is a complex mixture of calcium/magnesium salts of ghattic acids and mixed monosaccharides (CAS 9000-28-6); the water-soluble fraction has an approximate molecular weight of 12,000 Da.
Catalog No. 50-002-83838 Supplier Medchemexpress LLC Supplier No. HYW14551550G
Gum ghatti is a natural exudate polysaccharide supplied as a dry powder for research use. It serves as a biochemical reagent and excipient for assays, formulations, and other laboratory applications requiring a water-soluble gum. The material is a complex mixture of calcium/magnesium salts of ghattic acids and mixed monosaccharides (CAS 9000-28-6); the water-soluble fraction has an approximate molecular weight of 12,000 Da.