Learn More
Medchemexpress LLC Gum ghatti (Indian gum) | 9000-28-6 | 25 G
SDP
Catalog No. 5000283221
Encompass_Preferred
Gum ghatti (CAS 9000-28-6) is a natural, high-molecular-weight polysaccharide exudate from Anogeissus latifolia used as a biochemical reagent and formulation stabilizer in life-science research. It is water-soluble, forms viscous mucilage, and supports emulsification and film formation. Material is supplied for research use only.
  • Natural polysaccharide from Anogeissus latifolia.
  • High, polydisperse molecular weight supporting emulsification.
  • Water-soluble and forms viscous mucilage.
  • Functions as stabilizer, emulsifier, and film former in formulations.
  • Intended for research use only; not for therapeutic use.
Catalog No. 50-002-83221 Supplier Medchemexpress LLC Supplier No. HYW14551525G
Gum ghatti (CAS 9000-28-6) is a natural, high-molecular-weight polysaccharide exudate from Anogeissus latifolia used as a biochemical reagent and formulation stabilizer in life-science research. It is water-soluble, forms viscous mucilage, and supports emulsification and film formation. Material is supplied for research use only.
  • Natural polysaccharide from Anogeissus latifolia.
  • High, polydisperse molecular weight supporting emulsification.
  • Water-soluble and forms viscous mucilage.
  • Functions as stabilizer, emulsifier, and film former in formulations.
  • Intended for research use only; not for therapeutic use.