Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gum guaiac, MP Biomedicals™

Catalog No. ICN15729150 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15729150 50 g
ICN15729180 100 g
ICN15729190 500 g
ICN15729191 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15729150 Supplier MP Biomedicals, Inc Supplier No. 0215729150

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 9000-29-7

Specifications

Color Brown to Brown/Green
Quantity 50 g
Physical Form Powder
Chemical Name or Material Gum guaiac
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.