Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gum Rosin

Catalog No. 608951KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-895-1KG 1 kg
60-895-250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 60-895-1KG Supplier Sigma Aldrich Supplier No. 608951KG
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Gum rosin is an exudate gum, which is extracted from pine trees. It consists of abiatic acid, palaustric acid and neoabetic acid.Gum rosin has been used:for the preparation of testosome patchesto study its influence on the physical stability of concentrated oil in water emulsionsfor the preparation of rosin nanoparticles containing hydrocortisone (HC)

Specifications

CAS 8050-09-7
MDL Number MFCD00132205
Quantity 1 kg
Synonym Colophony; Rosin, gum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.