Learn More
Medchemexpress LLC Gum tragacanth | 9000-65-1 | MFCD00131255 | 840,000 g/mol | No single chemical formula (complex mixture of polysaccharides) | 25 G
SDP
Catalog No. 5000280859
Encompass_Preferred
Tragacanth gum is a natural anionic polysaccharide powder extracted from Astragalus species and supplied for laboratory research applications. It consists of high-molecular-weight polysaccharide fractions used as a stabilizer, emulsifier, binder, and functional additive in biochemical assays and materials synthesis.
  • Natural anionic polysaccharide powder.
  • High-molecular-weight polysaccharide composition.
  • Functions as an emulsifier and stabilizer in formulations.
  • Can improve cell adhesion in biological assays.
  • Exhibits nephroprotective and anticoagulant potential.
  • Acts as a reducing agent and stabilizer in nanoparticle synthesis.
  • Available in multiple pack sizes, including 25 g.
Catalog No. 50-002-80859 Supplier Medchemexpress LLC Supplier No. HYW25012125G
Tragacanth gum is a natural anionic polysaccharide powder extracted from Astragalus species and supplied for laboratory research applications. It consists of high-molecular-weight polysaccharide fractions used as a stabilizer, emulsifier, binder, and functional additive in biochemical assays and materials synthesis.
  • Natural anionic polysaccharide powder.
  • High-molecular-weight polysaccharide composition.
  • Functions as an emulsifier and stabilizer in formulations.
  • Can improve cell adhesion in biological assays.
  • Exhibits nephroprotective and anticoagulant potential.
  • Acts as a reducing agent and stabilizer in nanoparticle synthesis.
  • Available in multiple pack sizes, including 25 g.