Learn More
Cayman Chemical Guanabenz (hydrochloride), Purity: ≥98%, CAS: 23113-43-1, 100 mg
SDP
Catalog No. 502951091
Encompass_Preferred
An α2-adrenergic receptor agonist (effective concentrations 10-100 nM) with hypotensive effects; competes for imidazoline I2-binding sites in brown adipose tissue (Ki = 97 nM)
Catalog No. 50-295-1091 Supplier Cayman Chemical Supplier No. 10851100 MG
An α2-adrenergic receptor agonist (effective concentrations 10-100 nM) with hypotensive effects; competes for imidazoline I2-binding sites in brown adipose tissue (Ki = 97 nM)