Learn More
Cayman Chemical Guanabenz (hydrochloride), Purity: ≥98%, CAS: 23113-43-1, 50 mg
SDP
Catalog No. 502951367
Encompass_Preferred
An α2-adrenergic receptor agonist (effective concentrations 10-100 nM) with hypotensive effects; competes for imidazoline I2-binding sites in brown adipose tissue (Ki = 97 nM)
Catalog No. 50-295-1367 Supplier Cayman Chemical Supplier No. 1085150 MG
An α2-adrenergic receptor agonist (effective concentrations 10-100 nM) with hypotensive effects; competes for imidazoline I2-binding sites in brown adipose tissue (Ki = 97 nM)