Learn More
Medchemexpress LLC Gunagratinib | 2211082-53-8 | 98.5% | 423.47 g·mol⁻¹ | C22H25N5O4 | 5 MG
SDP
Catalog No. 5000262250
Encompass_Preferred
Gunagratinib is a covalent, orally active pan-FGFR inhibitor supplied as a purified research compound for preclinical studies. It irreversibly inhibits FGFR signaling through covalent binding and is provided in multiple small-scale pack sizes and formats for use in in vitro and in vivo research.
  • Covalent pan-FGFR inhibitor with irreversible binding.
  • Orally active compound suitable for preclinical studies.
  • High purity (98.5%) for research applications.
  • Molecular formula C22H25N5O4; molecular weight 423.47 g·mol⁻¹.
  • Available in small pack sizes (including 5 mg) as solid or DMSO solution.
  • Supplied for laboratory research use only.
Catalog No. 50-002-62250 Supplier Medchemexpress LLC Supplier No. HY1328175MG
Gunagratinib is a covalent, orally active pan-FGFR inhibitor supplied as a purified research compound for preclinical studies. It irreversibly inhibits FGFR signaling through covalent binding and is provided in multiple small-scale pack sizes and formats for use in in vitro and in vivo research.
  • Covalent pan-FGFR inhibitor with irreversible binding.
  • Orally active compound suitable for preclinical studies.
  • High purity (98.5%) for research applications.
  • Molecular formula C22H25N5O4; molecular weight 423.47 g·mol⁻¹.
  • Available in small pack sizes (including 5 mg) as solid or DMSO solution.
  • Supplied for laboratory research use only.