Learn More
Medchemexpress LLC Gunagratinib | 2211082-53-8 | 98.5% | C22H25N5O4 | 10MG
SDP
Catalog No. 5000258522
Encompass_Preferred
Gunagratinib is a research small molecule that irreversibly and covalently inhibits fibroblast growth factor receptors (FGFR). It is orally active in preclinical models and is used in cancer research and biochemical studies; the molecule also contains an alkyne handle that allows copper-catalyzed azide-alkyne cycloaddition (click chemistry) applications.
Catalog No. 50-002-58522 Supplier Medchemexpress LLC Supplier No. HY13281710MG
Gunagratinib is a research small molecule that irreversibly and covalently inhibits fibroblast growth factor receptors (FGFR). It is orally active in preclinical models and is used in cancer research and biochemical studies; the molecule also contains an alkyne handle that allows copper-catalyzed azide-alkyne cycloaddition (click chemistry) applications.