Learn More
Cayman Chemical GunosIn 5-dIphosphatesod 10mg
SDP
Catalog No. 502991186
Encompass_Preferred
A purine nucleotide; precursor of GTP; has been used to study GTPases; activates the SUR2B/Kir6.1 channel in HEK293T cells in a patch-clamp assay at 100 µM
Catalog No. 50-299-1186 Supplier Cayman Chemical Supplier No. 3170410 MG
A purine nucleotide; precursor of GTP; has been used to study GTPases; activates the SUR2B/Kir6.1 channel in HEK293T cells in a patch-clamp assay at 100 µM